Indigo carmine

Leverandør: Molekula
Warning

Synonymer: CI 73015, FD&C Blue #2, 5,5'-Indigodisulphonic acid, Disodium salt, Food blue No. 2, acid Blue 74, Sodium 3,3'-dioxo-[2,2'-biindolinylidene]-5,5'-disulphonate, FD and C Blue No. 2, Carmine Blue

M30726953
MOLEM30726953EA 720 NOK
MOLEM30726953
Indigo carmine
Indigo carmine
Formel: C₁₆H₈N₂Na₂O₈S₂
MW: 466.36 g/mol
Smeltepunkt: > 300 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00005723
CAS-nummer: 860-22-0
EINECS: 212-728-8
Merck Indeks: 13,04969

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR