Carbenicillin disodium salt

Leverandør: Molekula
Danger

Synonymer: NSC 111071, α-Carboxybenzylpenicillin disodium salt, Sodium (2S,5R,6R)-6-(2-carboxylato-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, 6-[(Carboxyphenylacetyl)-amino]-3,3-dimethyl-7-oxo-4-thia-1-aza-bicyclo[3.2.0]heptane-2-carboxylic Acid, 2Na

M32113067
MOLEM32113067EA 0 NOK
MOLEM32113067
Carbenicillin disodium salt
Carbenicillin disodium salt
Formel: C₁₇H₁₆N₂O₆S·2Na
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00077683
CAS-nummer: 4800-94-6
EINECS: 225-360-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR