Indol

Leverandør: Molekula
Danger

Synonymer: 1H-Indole

M32270590
MOLEM32270590EA 3725 NOK
MOLEM32270590
Indol
Indole
Formel: C₈H₇N
MW: 117,15 g/mol
Kokepunkt: 254…255 °C (1013 hPa)
Smeltepunkt: 50…51 °C
Tetthet: 1,22 g/cm³ (20 °C)
Flash punkt: 110 °C
MDL nummer: MFCD00005607
CAS-nummer: 120-72-9
EINECS: 204-420-7
FN: 2811
ADR: 6.1,III
Merck Indeks: 13,04985

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR