L(+)-Lithium lactate

Leverandør: Molekula

Synonymer: Lithium (S)-2-hydroxypropanoate, (S)-(+)-Lactic acid lithium salt, (S)-2-Hydroxypropionic acid Lithium salt, Lithium (S)-2-Hydroxypropionate, L(+)-Lactic acid lithium salt, (S)-(+)-2-Hydroxypropanoic acid lithium salt, (S)-(+)-2-Hydroxypropionic acid lithium salt, Lithium L-Lactate, L-Lactic acid Lithium salt

M33846384
MOLEM33846384EA 1675 NOK
MOLEM33846384
L(+)-Lithium lactate
L(+)-Lithium lactate
Formel: C₃H₅LiO₃
MW: 96.01 g/mol
Kokepunkt: 200
Smeltepunkt: 75
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00065512
CAS-nummer: 27848-80-2
EINECS: 248-692-5

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR