Bilirubin

Leverandør: Molekula

Synonymer: 3-(2-((3-(2-Carboxyethyl)-4-methyl-5-((Z)-(3-methyl-5-oxo-4-vinyl-1H-pyrrol-2(5H)-ylidene)methyl)-1H-pyrrol-2-yl)methyl)-4-methyl-5-((Z)-(4-methyl-5-oxo-3-vinyl-1H-pyrrol-2(5H)-ylidene)methyl)-1H-pyrrol-3-yl)propanoic acid

M43664818
MOLEM43664818EA 2200 NOK
MOLEM43664818
Bilirubin
Bilirubin
Formel: C₃₃H₃₆N₄O₆
MW: 584.67 g/mol
Smeltepunkt: > 360 °C
Tetthet: 1.31 g/cm³ (20 °C)
MDL nummer: MFCD00005499
CAS-nummer: 635-65-4
EINECS: 211-239-7

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR