Veratraldehyd

Leverandør: Molekula
Warning

Synonymer: 3,4-Dimethoxybenzaldehyde

M43971872
MOLEM43971872EA 2200 NOK
MOLEM43971872
Veratraldehyd
Veratraldehyd
Formel: C₉H₁₀O₃
MW: 166,18 g/mol
Kokepunkt: 281 °C (1013 hPa)
Smeltepunkt: 40…43 °C
Flash punkt: >100 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003363
CAS-nummer: 120-14-9
EINECS: 204-373-2
Merck Indeks: 12,10084

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR