Tris(dimethylamino)phosphine

Leverandør: Molekula

M44821458
MOLEM44821458EA 1900 NOK
MOLEM44821458
Tris(dimethylamino)phosphine
Tris(dimethylamino)phosphine
Formel: C₆H₁₈N₃P
Kokepunkt: 150 °C (1013 hPa)
Smeltepunkt: –44 °C
Tetthet: 0,8962 g/cm³ (20 °C)
MDL nummer: MFCD00008301
CAS-nummer: 1608-26-0
EINECS: 216-534-4

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR