Cresol red sodium salt

Leverandør: Molekula

Synonymer: Cresol Red sodium, Sodium 4-(3-(4-hydroxy-3-methylphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)-2-methylphenolate

M56421347
MOLEM56421347EA 1200 NOK
MOLEM56421347
Cresol red sodium salt
Cresol red sodium salt
Formel: C₂₁H₁₇NaO₅S
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00001618
CAS-nummer: 62625-29-0
EINECS: 263-654-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR