(2,4-Diklorofenoksy)eddiksyre

Leverandør: Molekula
Danger

Synonymer: 2,4-D

M63113303
MOLEM63113303EA 1050 NOK
MOLEM63113303
(2,4-Diklorofenoksy)eddiksyre
(2,4-Diklorofenoksy)eddiksyre
Formel: (C₆H₃Cl₂)OCH₂COOH
MW: 221.04 g/mol
Kokepunkt: 346
Smeltepunkt: 137…139 °C
Tetthet: 1.563 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004300
CAS-nummer: 94-75-7
EINECS: 202-361-1
Merck Indeks: 13,02825

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR