2-Aminopyridin

Leverandør: Molekula

Synonymer: 2-Pyridylamine, Pyridin-2-amine

M71719173
MOLEM71719173EA 1900 NOK
MOLEM71719173
2-Aminopyridin
2-Aminopyridine
Formel: C₅H₆N₂
MW: 94,12 g/mol
Kokepunkt: 210,6 °C (1013 hPa)
Smeltepunkt: 50…52 °C
Tetthet: 1,24 g/cm³ (20 °C)
Flash punkt: 92 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00006312
CAS-nummer: 504-29-0
EINECS: 207-988-4
FN: 2671
ADR: 6.1,II
Merck Indeks: 13,00472

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR