1,2-Difluorbensen

Leverandør: Molekula

M71909834
MOLEM71909834EA 1900 NOK
MOLEM71909834
1,2-Difluorbensen
1,2-Difluorbensen
Formel: C₆H₄F₂
MW: 114.09 g/mol
Kokepunkt: 92
Smeltepunkt: –34 °C
Tetthet: 1.15 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00000284
CAS-nummer: 367-11-3
EINECS: 206-680-7
FN: 1993
ADR: 3 II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR