4-Metoksysalisylsyre

Leverandør: Molekula
Warning

Synonymer: 2-Hydroxy-p-anisic acid, 2-Hydroxy-4-methoxybenzoic acid

M74022815
MOLEM74022815EA 3450 NOK
MOLEM74022815
4-Metoksysalisylsyre
4-Metoksysalisylsyre
Formel: C₈H₈O₄
MW: 168,15 g/mol
Kokepunkt: 327 °C (1013 hPa)
Smeltepunkt: 191 °C
MDL nummer: MFCD00002450
CAS-nummer: 2237-36-7
EINECS: 218-801-0

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR