5-Aminonicotinic acid

Leverandør: Molekula

Synonymer: 5-Aminopyridine-3-carboxylic acid, 5-Amino-3-pyridinecarboxylic acid

M75880604
MOLEM75880604EA 1850 NOK
MOLEM75880604
5-Aminonicotinic acid
5-Aminonicotinic acid
Formel: C₆H₆N₂O₂
MW: 138,13 g/mol
MDL nummer: MFCD00129116
CAS-nummer: 24242-19-1

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR