di-Ammoniumhydrogencitrat

Leverandør: Molekula
Warning

Synonymer: Citric acid diammonium salt, Diammonium hydrogen 2-hydroxypropane-1,2,3-tricarboxylate, Ammonium citrate dibasic

M77018759
MOLEM77018759EA 1550 NOK
MOLEM77018759
di-Ammoniumhydrogencitrat
di-Ammoniumhydrogencitrat
Formel: HOC(CO₂H)(CH₂CO₂NH₄)₂
MW: 226.19 g/mol
Tetthet: 1.483 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00013068
CAS-nummer: 3012-65-5
EINECS: 221-146-3
Merck Indeks: 13,00513

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR