3-Methyl-1-phenyl-2-pyrazolin-5-one

Leverandør: Molekula

M79333761
MOLEM79333761EA 1150 NOK
MOLEM79333761
3-Methyl-1-phenyl-2-pyrazolin-5-one
3-Methyl-1-phenyl-2-pyrazolin-5-one
Formel: C₁₀H₁₀N₂O
MDL nummer: MFCD00003138
CAS-nummer: 89-25-8
EINECS: 201-891-0

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR