Fenol rødt

Leverandør: Molekula

Synonymer: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

M86776424
MOLEM86776424EA 1850 NOK
MOLEM86776424
Fenol rødt
Fenol rødt
Formel: C₁₉H₁₄O₅S
MW: 354.38 g/mol
Smeltepunkt: > 300 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003552
CAS-nummer: 143-74-8
EINECS: 205-609-7
Merck Indeks: 13,07329

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR