SFC-100FLED Corded Stereo Microscope

Leverandør: MOTIC
1100100800653
MOTI1100100800653EA 2550 NOK
MOTI1100100800653
SFC-100FLED Corded Stereo Microscope
Mikroskop
The perfect entry into the world of microscopy; robust, easy to handle, and affordable: The Motic SFC100 Series offers the idea instrument for a first step into the 'world of small things'.

No plastics, but efficient optics 100% made of coated glass, this is the way to crisp and clear images for a long-term usage in classroom or for leisure activities. The SFC100 is a classic microscope with a classic performance.

Inclination: 45° inclined, 360° rotating
Objective classification: Achromatic, DIN
Objective mounting thread: W ⁴/₅×¹/₃₆ " (RMS standard)
Diaphragm: Iris diaphragm

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Kontrastteknikk Lysfelt
Okularer WF 10×/18 mm med diopter
Objektivrevolver 3 posisjoner
Objektiver 4× 0,10 (WD 15,9 mm), 10× 0,25 (WD 17,4 mm), 40× 0,65/S (WD 0,5 mm)
Plattform Plain stage with sample clips, 120×110 mm
Kondensor Built-in condenser lens N.A. 0,65
Belysning LED 3,5 V/70 mW
Stativ Upright
Elektronisk 220 - 240 V (CE)
B×D×H 175×133×358 mm
Vekt 2,7 kg
Optisk system Finite optical system, 160 mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR