Metallurgical microscopes, BA310 MET
Leverandør: MOTIC
1100100402042 1100100402141 1100100402501 1100100402511 1100100402603 1101000201982 1101001704392 1101001704402 1101001704512 1101001704601 1101001704611 1101001901609 1101002402431 1100100402521
630-2689EA 58100 NOK
630-2689 630-2690 MOTI1100100402501 MOTI1100100402511 630-2605 MOTI1101000201982 MOTI1101001704392 MOTI1101001704402 MOTI1101001704512 MOTI1101001704601 MOTI1101001704611 MOTI1101001901609 630-2044 MOTI1100100402521
Metallurgical microscopes, BA310 MET
Mikroskop
These metallurgical microscopes are designed for viewing opaque materials such as minerals and metals.
  • High quality glass objectives enabling improved contrast and image quality
  • Flexible focus dedicated to large samples
  • Various lighting options for incident and/or transmitted light
Suitable for educational environments of engineering or material professionals.
NOTE: For increased illumination, replace the halogen bulb by an LED module.