LED illuminator, VI-LED vertical illuminator
Leverandør: MOTIC
1101010100151
MOTI1101010100151EA 4625 NOK
MOTI1101010100151
LED illuminator, VI-LED vertical illuminator
Lys LED-belysnings
VI-LED vertical illuminator enables on-axis observation and documentation, and is specially designed for Motic SMZ-161 and SMZ-171. VI-LED directs light from the 3 W LED down through the objective onto the specimen. It is powered by the switching power supply for worldwide use (100 to 240 V). Ideal for the observation of particularly smooth, specular surfaces and highly reflective specimens such as integrated circuits, semi-conductor wafers, polished metal parts, solder balls, or magnetic recording heads.
  • True on-axis observation of high-resolution, high-contrast, shadow-free images