Extraction systems and accessories for industrial welding
Leverandør: asecos
10170 10167 10169 10171 10172 10173 10174 12868 10175 10182 10184
MSGA10170EA 16600 NOK
MSGA10170 MSGA10167 MSGA10169 MSGA10171 MSGA10172 MSGA10173 MSGA10174 MSGA12868 MSGA10175 MSGA10182 MSGA10184
Extraction systems and accessories for industrial welding
Avtrekksekstraktor
These extraction systems are ideal for use in narrow places such as welding booths or at bank work. They have externally mounted brackets for natural, straight run of the exhaust air.
  • Integrated throttle
  • Extremely light and stable position
  • Gas spring for balancing the dead weight
  • Low pressure drop for effective extraction