Safety storage cabinets, type 30, E-PSM-UF
Leverandør: asecos
30630
MSGA30630EA 0 NOK
MSGA30630
Safety storage cabinets, type 30, E-PSM-UF
Møbler Kjemikalieskap
The cabinets are height adjustable with galvanised tray with loading capacity of 50 kg.
  • Sturdy sheet-steel construction
  • Connection with a technical exhaust system
Sertifiseringer: EN14470-1
Bestillingsinformasjon: Interior equipment packages need to be ordered separately (not included). Please check the website or contact Avantor for further information.