Relay Module
Leverandør: asecos
38766
MSGA38766EA 3975 NOK
MSGA38766
Relay Module
Møbler Kjemikalieskap
By choosing this option, you are opting for alarm differentiation instead of a collective alarm.
These signals are passed on to a central control centre via potential-free alarm contacts. With the clear differentiation of the alarm conditions, you can initiate optimally coordinated measures.
The relay module is suited for the PRO models of the IONLINE.