Wide Storage Tower
Leverandør: MULTIROIR
104461
MULC104461EA 2250 NOK
MULC104461
Wide Storage Tower
Møbler Kjemikalieskap
This wide three-drawer polypropylene tower is ideal for storing various materials, whether they are consumables or stationery.
  • 3 transparent drawers, each with a 30 L capacity
  • 4 self-assembly multidirectional wheels
  • Lid reinforced with a metal bar, capable of supporting up to 100 kg
  • Drawers with large handles
  • Anti-drop stops on drawers
Made entirely of polypropylene, this storage tower allows for the storage of various products. Additionally, it is resistant to temperature variations, chemicals, and impacts. Therefore, it can be placed in bothMore Product Information