Storage Unit with Six Drawer Trays, PM
Leverandør: MULTIROIR
31104
MULC31104EA 2700 NOK
MULC31104
Storage Unit with Six Drawer Trays, PM
Møbler Kjemikalieskap
Discover the block six drawers model PM, ideal for use in the laboratory.
  • Six drawer trays in mixed colours
  • Made from impact-resistant polystyrene
  • Integrated handle on each tray
  • Drawer trays can be stacked on top of each other
The small 6-drawer storage unit is entirely made of high-impact polystyrene. The drawers are removable and can be used as preparation trays. They can also be safely stacked on top of each other.
Lightweight and durable, this drawer unit is suitable for laboratory use for preparing elements or storing materials.