Naphthol AS-BI phosphate ≥98% (by HPLC), Technical Grade, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY


M-Clarity™ quality level = MQ200

N2125-100MG N2125-1G N2125-500MG N2125-5G
N2125-100MGEA 957 NOK
N2125-100MG N2125-1G N2125-500MG N2125-5G
Naphthol AS-BI phosphate ≥98% (by HPLC), Technical Grade, Sigma-Aldrich®
Naphthol AS-BI phosphate

Substrate for the histochemical demonstration of acid and alkaline phosphatase. Substrate for the fluorometric assay of acid and alkaline phosphatases. Demonstration of acid phosphatase in paraffin embedded tissue. Cytofluorometric assay of acid phosphatase in single living cells.


  • Suitable for haematology
  • Suitable for histology

Formel: C₁₈H₁₅BrNO₆P
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00004058
CAS-nummer: 1919-91-1
EINECS: 217-645-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96cafb60079b0bd0a25c6e289e787950690ecd94&vid=69276bc3177bccaa86ee3353c2578faea6bb9e1d&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR