Naphthol AS-MX phosphate disodium salt, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY


M-Clarity™ quality level = MQ200

N5000-100MG N5000-1G N5000-500MG
N5000-100MGEA 468 NOK
N5000-100MG N5000-1G N5000-500MG
Naphthol AS-MX phosphate disodium salt, Sigma-Aldrich®
Naphthol AS-MX phosphate disodium salt

Naphthol AS-MX phosphate disodium salt,phosphatase substrate

Formel: C₁₉H₁₆NNa₂O₅P
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00069549
CAS-nummer: 96189-12-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32505f4bad1f69ee9b3ece24518842ec141a2406&vid=d385c48ce0aede1afdd9b6d72016b24ed9ffc336&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance (Colour) White to Off-White
Appearance (Form) Powder
Solubility (Colour) Faint Yellow
Solubility (Turbidity)
100 mg/ml in H2O
Clear to Slightly Hazy
Infrared spectrum Conforms to Structure
Water (by Karl Fischer)
(10 mol/mol)
≤31 %
Carbon (anhydrous) 53.5 - 56.6 %
Nitrogen (anhydrous) 3.0 - 3.6 %
Phosphorus (P) anhydrous 7.1 - 7.7 %
Sodium (Na) anhydrous 10.0 - 12.0 %
Purity (HPLC) ≥99 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR