Nøytralrødt 90% (dye content), Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY


M-Clarity™ quality level = MQ200

N7005-25G N7005-5G N7005-1G
N7005-25GEA 7770 NOK
N7005-25G N7005-5G N7005-1G
Nøytralrødt 90% (dye content), Sigma-Aldrich®
Nøytralrødt

Neutral red (C.I. 50040) is a protonated diaminophenazine dye. It is aminodimethylaminomethylphenazinehydrochloride, C15H17N4Cl.


Neutral red has multiple staining applications that include its use as a nuclear stain, counterstain in the Gram procedure, either alone, or as a component of the Twort stain (a fast green-neutral red mixture). Neutral red staining is also a viability assessment method used for the detection of viable yeast cells in dandruff samples and of mussel larvae. Neutral red is also recommended for use in viability assessment as it is selectively absorbed by lysosomes and other acidic organelles in cells. It is also used for supravital staining of living blood cells. Useful as an indicator for preparing neutral red paper, and as a biological stain. Neutral red is a widely used cationic fluorochrome that is known to accumulate in the Golgi complex and nuclei after first staining lysosomes.

Formel: C₁₅H₁₇ClN₄
Smeltepunkt: 290 °C (dec., lit.)
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00012651
CAS-nummer: 553-24-2
EINECS: 209-035-8
Merck Indeks: 12,06571
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6287f6f6bcfd94a7d607e01e26d64e6223b3091&vid=74c977d74889ce8bd72a9b90e9ce07615ef7fb94&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance (Colour) Brown to Black may have red or green sheen
Appearance (Form) Powder
Solubility (Colour), 10 mg/ml, H₂O Dark Red
Infrared spectrum Conforms to Structure
Dye Content (Calculated based on UV spectral assay) ≥90%
Recommended Retest Period 4 years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR