VWR® Ready Ladder™ DNA Ladder, 50 bp
Leverandør: VWR Chemicals
N746-100RXNEA 2425 NOK
N746-100RXN
VWR® Ready Ladder™ DNA Ladder, 50 bp
Elektroforese reagenser Molekylvekt markører
Ladder with 10 fragments ranging from 50 to 500 bp for easy band identification. Ready to use formula pre-mixed with loading dye.