Natriumhydrogensulfitt, Reagent Grade

Leverandør: VWR Chemicals
Danger

Synonymer: Sodium hydrogen sulphite

N822-500GEA 3650 NOK
N822-500G
Natriumhydrogensulfitt, Reagent Grade
Natriumhydrogensulfitt
Formel: HNaO₃S
MW: 104.06 g/mol
Smeltepunkt: 300 °C
Tetthet: 2.476 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00167602
CAS-nummer: 7631-90-5
EINECS: 231-548-0
FN: 2693
ADR: 8 III
Merck Indeks: 12,08731

Bestill nå

Spesifikasjon Testresultater

Chloride Content <0.02%
Heavy Metals (as Pb) <0.001%
Insoluble Matter <0.005%
Iron <0.002%
Titration by Na₂SO₃ (%SO₂) >58.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR