Charging Devices with and without Cooling Fan for N-Series
Leverandør: NABERTHERM
631000528
NABE631000528EA 0 NOK
NABE631000528
Charging Devices with and without Cooling Fan for N-Series
Ovner Høytemperatursovner
For charging larger workpieces and annealing boxes.
  • 4 casters, freely movable
  • Equipped with a rack at working height for temporary storage
  • Fixing lock for annealing bags (CWK)
  • CWK version with cooling fan (0.2 kW, 230 V)