Tanker, sylindriske, med stoppekran og deksel, Nalgene®
Leverandør: Thermo Fisher Scientific
11102-0005 11102-0007 11102-0010 11102-0015 11102-0030 11102-0055
NALG11102-0005EA 4900 NOK
NALG11102-0005 NALG11102-0007 NALG11102-0010 NALG11102-0015 NALG11102-0030 NALG11102-0055
Tanker, sylindriske, med stoppekran og deksel, Nalgene®
Store beholdere Store beholdere, sylindriske
These heavy-duty cylindrical LLDPE tanks with cover and spigot can withstand most acid, alcohols and bases. Tanks used at elevated temperatures or with high specific-gravity liquids should be externally supported.
  • Economical
  • Impact- and abrasion-resistant
  • Not autoclavable
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact
Miljøsertifikat NAR Certificate of Retirement