Flasker, til emballering, smal munnet, rund, PPCO, med skrukork, Nalgene®
Leverandør: Thermo Fisher Scientific
312087-0001 312087-0002 312087-0004 312087-0008 312087-0016 312087-0032
NALG312087-0001EA 18800 NOK
NALG312087-0001 NALG312087-0002 NALG312087-0004 NALG312087-0008 NALG312087-0016 NALG312087-0032
Flasker, til emballering, smal munnet, rund, PPCO, med skrukork, Nalgene®
Flasker Plastflasker
PPCO, translucent, with PP screw cap.
  • Excellent clarity
  • Impact and chemical resistance
  • Leakproof
  • Autoclavable
These bottles have a robust construction for packaging use.
Certifications: Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Packaging: Bottles and closures are packed in separate polybags within the same carton.
Order Now