Flasker, smal munn, HDPE, fluorbehandlet, med skrulokk, rund, Nalgene®
Leverandør: Thermo Fisher Scientific
312097-0004 312097-0008 312097-0016 312097-0032
NALG312097-0004EA 23575 NOK
NALG312097-0004 NALG312097-0008 NALG312097-0016 NALG312097-0032
Flasker, smal munn, HDPE, fluorbehandlet, med skrulokk, rund, Nalgene®
Flasker Plastflasker
HDPE, fluorinated with PP, fluorinated screw cap.
  • Fluorinated bottles and carboys can be used for most acids, alkalis and aggressive organic solvents
  • Round shape, non sterile
  • Heavy-duty walls are safe, durable and resistant to splitting and punctures
  • Fluorocarbon surfaces inside and out
  • Leakproof
Fluorinating these HDPE bottles and carboys provides a cost effective alternative to costly fluoropolymer containers.
Manufactured from Nalgene™ for resistance to permeation, paneling, odor emission, flavor and fragrance loss. Provide superior recepticalsMore Product Information