Flasker, smal hals, rund, HDPE, Boston type, med gjenger, Nalgene®
Leverandør: Thermo Fisher Scientific
362002-0002 362002-0004 362002-0001 362002-0008 362002-0016
NALG362002-0002EA 16425 NOK
NALG362002-0002 NALG362002-0004 NALG362002-0001 NALG362002-0008 NALG362002-0016
Flasker, smal hals, rund, HDPE, Boston type, med gjenger, Nalgene®
Flasker Plastflasker
HDPE, translucent with PP screw cap.
  • Good chemical resistance
  • Heat resistant from −100 to +120 °C
  • Leakproof
Ideal for storing, packaging and transporting liquids. Robust, strong and stable.
Sertifiseringer: Manufactured in compliance with ISO 13485:2016 quality requirements. Diagnostic Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.