Biotainer® Replacement Screw Caps for Bottles, InVitro™
Leverandør: Thermo Fisher Scientific
362515-0480
NALG362515-0480EA 58250 NOK
NALG362515-0480
Biotainer® Replacement Screw Caps for Bottles, InVitro™
Lokk Flaskelokk
Protect the contents in your Thermo Scientific™ Biotainer™ bottles and carboys. Thermo Scientific™ Nalgene™ sterile polypropylene biotainer replacement closures are for 1, 2, 5, 10 and 20 L polycarbonate biotainers.
  • Designed to fit only nalgene bottles to provide a guaranteed leakproof system
  • Polypropylene with silicone liner
  • Closure size: 48 mm
  • Packaged individually: 1 per bag, 6 bags or 50 per case, 300 in a case