Nalgene® Bottles, Narrow Neck, Round, LDPE, Low Particulate/Low Metals, wiith Screw Caps
Leverandør: Thermo Fisher Scientific
382003-0004 382003-0008 382003-0032 382003-0016
NALG382003-0004EA 3200 NOK
NALG382003-0004 NALG382003-0008 NALG382003-0032 NALG382003-0016
Nalgene® Bottles, Narrow Neck, Round, LDPE, Low Particulate/Low Metals, wiith Screw Caps
Flasker Plastflasker
LDPE, translucent, with PP screw cap.
  • Certified to particulate and trace metals levels
  • Excellent chemical resistance to most acids, bases and alcohols
  • Guaranteed leakproof
  • Non sterile
Metals certified to microlitre (ppb) levels <0,20 Hg, <0,5 Be, <1,0 As, Cd, Pb, <2,0 Sb, Se, <5,0 Ag, Co, Cr, Cu, Mn, Tl, V, <10 Ba, Ni, Zn, <75 Al, <100 Ca, Fe, K, Na. Each bottle is double bagged under Class 10 laminar flow hoods inside a Class 100 clean room. Excellent for ICP-MS reagent and standard storage. Product includes a certificate ofMore Product Information