Horisontale stativ, for flerbrønnsplater, Nalgene®
Leverandør: Thermo Fisher Scientific
5039-0048
NALG5039-0048EA 6950 NOK
NALG5039-0048
Horisontale stativ, for flerbrønnsplater, Nalgene®
Stativer for rør Mikroplatestativer
Stainless steel.
  • Racks fit 1,0 ml cryobank cryogenic storage vials
  • Horizontal design fits most refrigerators and freezers
  • Retainer clip on each compartment holds plates securely in place
  • Handles on tops and sides for easy transport
  • Label holder for easy identification of contents of rack
Securely store and transport samples in microplate, deep-well plates and cryobank racks using these horizontal storage racks.