Pipette box holders, Nalgene®
Leverandør: Thermo Fisher Scientific
5810-0002 5810-0004
NALG5810-0002EA 0 NOK
NALG5810-0002 NALG5810-0004
Pipette box holders, Nalgene®
Pipettestativ
The acrylic pipette box holder (two compartments) and the angled pipette holder (four compartments) accommodate bags, boxes, pipette canisters or 1- to 100- ml individually-wrapped serological pipettes.
  • Compartments sized to fit one box of pipettes (14,6 cm wide opening)
  • Storage area for rubber bulbs
  • Divider can be removed to form one large storage area
  • Can be mounted under the shelf with two brackets
  • Angled pipette holder for easy access; prevents pipettes from sliding out
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact
Miljøsertifikat NAR Certificate of Retirement