Slangeklemmer, Nalgene®
Leverandør: Thermo Fisher Scientific
6165-0002
NALG6165-0002EA 410 NOK
NALG6165-0002
Slangeklemmer, Nalgene®
Klemmer Slangeklemmer
This pinch clamp made of PP enables precise, one-hand regulation of fluid flow - from completely open to restricted flow to completely closed.
  • Plastic clamp will not rust, corrode or deform
  • Molded in one piece with no sharp edges
  • Accommodates a wide range of tubing