Dropping pipettes, Nalgene®
Leverandør: Thermo Fisher Scientific
6219-0068
NALG6219-0068EA 925 NOK
NALG6219-0068
Dropping pipettes, Nalgene®
Pipetter Pasteurpipetter og overføringspipetter
LDPE. Moulded in one piece. Non wetting material, tapered tip, and bellows-type bulb provide excellent drop formation.
  • Delivers 30 to 35 drops per ml
  • Safe and economical
  • Easy to clean for repeatable performance
  • Chemically compatible with most acids, bases and alcohols
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact
Miljøsertifikat IRECS; ISO 14001 (Certificate Registration No. EMS 752603)