Stopcocks, Needle-Type, Nalgene®
Leverandør: Thermo Fisher Scientific
6421-0010
NALG6421-0010EA 1100 NOK
NALG6421-0010
Stopcocks, Needle-Type, Nalgene®
Haner
PP, white.
  • Excellent chemical resistance
  • Excellent temperature resistance
Ensure safe and efficient liquid dispensing from tanks by using Nalgene™ spigots for storage tanks. Spigots are for use with Nalgene™ tanks up to 100 gal. with factory installed threaded boss.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact
Miljøsertifikat NAR Certificate of Retirement