Stopcocks, needle, Nalgene®
Leverandør: Thermo Fisher Scientific
96423-0100
NALG96423-0100EA 4275 NOK
NALG96423-0100
Stopcocks, needle, Nalgene®
Haner
Spigot is installed on a threaded HDPE boss which is welded onto the tank at the factory.
  • Boss fits only Nalgener tanks
  • Spigot Material: Polypropylene
  • Accepts ⁵/₈" inner Ø tubing
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact
Miljøsertifikat NAR Certificate of Retirement