Flasker, Valideringsflasker, Nalgene®
Leverandør: Thermo Fisher Scientific
DS2127-0250 DS2127-0030 DS2127-2000
NALGDS2127-0250EA 9125 NOK
NALGDS2127-0250 NALGDS2127-0030 NALGDS2127-2000
Flasker, Valideringsflasker, Nalgene®
Flasker Plastflasker
PC, clear, with PP screw cap.
  • Thick walls
  • Non-toxic
  • Fully autoclavable
Validation bottles provide material compatibility validation when scaling up production to vessels of larger sizes.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact
Miljøsertifikat NAR Certificate of Retirement