Pour spout, Nalgene®
Leverandør: Thermo Fisher Scientific
DS2176-0384
NALGDS2176-0384EA 1700 NOK
NALGDS2176-0384
Pour spout, Nalgene®
Lokk Lokk til dråpeflasker
PP, translucent. with polyethylene insert.
  • Used for all Nalgene bottles with 38-430 neck finish
  • Good chemical resistance
  • Good temperature resistance
Allows easy one-handed liquid dispensing without leaving bottles completely open to the environment.