Automated cell counter, ADAM-CellT
Leverandør: NANOENTEK
ADAM-CELLT-EU
NANOADAM-CELLT-EUEA 0 NOK
NANOADAM-CELLT-EU
Automated cell counter, ADAM-CellT
Celletellere Elektroniske celletellere
ADAM-CellT is an automated cell counter for cGMP. It takes only 25 seconds for ADAM-CellT to present results. ADAM-CellT has an easy-to-use built-in tablet and complies with 21 CFR part 11. When 21 CFR part 11 mode is activated, data cannot be modified by any user.
  • Automated image analysis
  • Accurate result
  • Sensitve CMOS detection and precise automatic stage
  • Auto focusing
  • Cell therapy quality control
To be used with 4 channel Accuchip slides.
Sertifiseringer: Compliance with 21 CFR part 11.