EVE™ HT Counting Kit
Leverandør: NANOENTEK
EVH-020
NANOEVH-020EA 15000 NOK
NANOEVH-020
EVE™ HT Counting Kit
Objektglass Tellekammer
EVETM HT counting kit consists of items necessary for counting cells with EVETM HT all in one package.
  • Counting plate has 48 channels of which 48 different samples can be counted at once
  • Only 10 μl of sample is required for each channel and yet high accuracy is provided
  • Measuring range detectable: 1×10E4 - 2×10E7 cells/ml and optimal: 1×10E5 - 1×10E7 cells/ml
Trypan blue staining solution dyes non-viable cells in blue for viability. It is an azo dye that is only applied to dead cells to help distinguish live and dead cells.
Using mix well plate,More Product Information