Sampling spoons, sterile
Leverandør: NASCO FORT ATKINSON
B01041AWA
NASCB01041AWAEA 970 NOK
NASCB01041AWA
Sampling spoons, sterile
Skjeer
PS, white. These gas sterilised spoons are ideal for collecting samples.
  • Made of sturdy, durable PS
  • Ideal for use with Whirl-Pak® bags
The procedure for contamination-free use is to open the bag, push one spoon up, pull it out, and reclose the bag.
Forpakning: 20 per bag, 100 per box.