Sludge nabber
Leverandør: NASCO FORT ATKINSON
B01229WA
NASCB01229WAEA 3750 NOK
NASCB01229WA
Sludge nabber
Prøvetakere Væskeprøvetakere
Sample bottles of various sizes are held via an adjustable locking device.
  • Makes sample collection of liquids fast and easy
  • Flexible with adjustable aluminum pole
  • PU holder with a plastic snapper ring
Suitable for numerous requirements including chlorine in solution – Can also be used for BOD.
Leveringsinformasjon: Supplied with a 500 ml PE bottle.