PIG® HazMat chemical loose absorbent
Leverandør: NEW PIG
HA8010
NEWPHA8010EA 1600 NOK
NEWPHA8010
PIG® HazMat chemical loose absorbent
Materialer for håndtering av søl
Loose absorbent can be sprinkled onto spills or spread into tight spaces for versatile clean-up of acids, caustics and unknown liquids.
  • Absorbs the widest range of acids, caustics and unknown liquids, even those with high concentrations like 98% sulfuric acid and 30% sodium hydroxide
  • Chemical-resistant pulp won't degrade or cause a dangerous reaction upon contact with corrosive spills
  • Pink colour is easily distinguishable to assure workers use the correct pulp during a spill emergency
  • Can be incinerated after use to reduce waste
  • Weight: 2,3 kg per bag