Refill spill kits, in extra-large response cart
Leverandør: NEW PIG
KITR204
NEWPKITR204EA 19350 NOK
NEWPKITR204
Refill spill kits, in extra-large response cart
Materialer for håndtering av søl
These refills are specially used for oils, coolants, solvents and water. Packed to refill the PIG Spill Kit in Extra-Large Response Chest or use as an economical boxed spill kit. This spill kit features an absorbents proven for rapid response.
  • PIG® Blue socks stop spreading spills; PIG® mat and pillows absorb; wipess clean surfaces and hands
  • Tamperproof seals help prevent pilfering of spill response supplies
  • Four easy-access compartments hold supplies to handle large spills
Kit includes 18× socks (80×300 mm), 55× socks (80×1220 mm), 150× heavy weightMore Product Information